Learn More
SPARC Rat anti-Mouse, Janelia Fluor 646, Clone: RM0130-5F41, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP122507JF646
Catalog No. NBP12507646
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
SPARC Monoclonal antibody specifically detects SPARC in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
SPARC
RM0130-5F41
Immunohistochemistry, Immunohistochemistry-Frozen
Purified
SPARC
Mouse recombinant protein of SPARC
Protein G purified
Biologically Active Proteins, Cancer, Cell Cycle and Replication
Primary
6678
Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin)
Rat
IgG2
0.1mL
Store at 4°C in the dark.
Monoclonal
Mouse