Learn More
Abnova™ SPARCL1 Recombinant Protein
Human SPARCL1 full-length ORF recombinant protein with GST-tag at N-terminal.
$463.00 - $713.00
Specifications
Cross Reactivity | Human |
---|---|
Format | Solution |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
For Use With (Application) | Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) |
Molecular Weight | 96.91 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-011-247
|
Abnova™
H00008404P0110 |
10 ug |
Each for $463.00
|
|
89-011-248
|
Abnova™
H00008404P0125 |
25 ug |
Each for $713.00
|
|
Description
- Theoretical MW (kDa): 96.91
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: PTNARLLSDHSKPTAETVAPDNTAIPSLRAEDEENEKETAVSTEDDSHHKAEKSSVLKSKEESHEQSAEQGKSSSQELGLKDQEDSDGDLSVNLEYAPTEGTLDIKEDMSEPQEKKLSENTDFLAPGVSSFTDSNQQESITKREENQEQPRNYSHHQLNRSSKHSQGLRDQGNQEQDPNISNGEEEEEKEPGEVGTHNDNQERKTELPREHANSKQEEDNTQSDDILEESDQPTQVSKMQEDEFDQGNQEQEDNSNAEMEEENASNVNKHIQETEWQSQEGKTGLEAISNHKETEEKTVSEALLMEPTDDGNTTPRNHGVDDDGDDDGDDGGTDGPRHSASDDYFIPSQAFLEAERAQSIAYHLKIEEQREKVHENENIGTTEPGEHQEAKKAENSSNEEETSSEGNMRVHAVDSCMSFQCKRGHICKADQQGKPHCVCQDPVTCPPTKPLDQVCGTDNQTYASSCHLFATKCRLEGTKKGHQLQLDYFGACKSIPTCTDFEVIQFPLRMRDWLKNILMQLYEANSEHAGYLNEKQRNKVKKIYLDEKRLLAGDHPIDLLLRDFKKNYHMYVYPVHWQFSELDQHPMDRVLTHSELAPLRASLVPMEHCITRFFEECDPNKDKHITLKEWGHCFGIKEEDIDENLLF
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Human | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
96.91 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Yes | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Solution | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
SPARCL1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.