Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spastin Mouse anti-Human, Mouse, Rat, Clone: Sp 6C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30784220UG

 View more versions of this product

Catalog No. NB123158


Add to cart

Description

Description

Spastin Monoclonal antibody specifically detects Spastin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Spastin
Sp 6C6
Unconjugated
10 mM PBS with 0.05% BSA
ADPSPSpastic paraplegia 4 protein, FSP2EC 3.6.4.3, KIAA1083EC 3.6.1.3, spastic paraplegia 4 (autosomal dominant; spastin), spastin, SPG4
Recombinant full-length human Spastin protein.
Protein A or G purified
RUO
Primary
6683
Western Blot, Immunofluorescence
0.2 mg/ml
Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Purified
Mouse
IgG2a Kappa
20 μg
Store at 4°C.
Monoclonal
Human, Mouse, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit