Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spastin Mouse anti-Human, Mouse, Rat, Clone: Sp 6C6, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Spastin
Clone Sp 6C6
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Western Blot, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123157
View Documents
Novus Biologicals
NBP30784210UG
100 μg
Each for $352.89
Add to cart
 
NB123158
View Documents
Novus Biologicals
NBP30784220UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Spastin Monoclonal antibody specifically detects Spastin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Spastin
0.2 mg/ml
Western Blot, Immunofluorescence
Purified
RUO
10 mM PBS with 0.05% BSA
6683
IgG2a Kappa
Primary
Store at 4°C.
Sp 6C6
Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Human, Mouse, Rat
ADPSPSpastic paraplegia 4 protein, FSP2EC 3.6.4.3, KIAA1083EC 3.6.1.3, spastic paraplegia 4 (autosomal dominant; spastin), spastin, SPG4
Recombinant full-length human Spastin protein.
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit