Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA2 Mouse anti-Human, Janelia Fluor 646, Clone: Z1P1F10&asterisk;F12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250304JF646
Description
SPATA2 Monoclonal specifically detects SPATA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
SPATA2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SPATA2 | |
Peptide Sequence - KSTQLSHLVYR | |
0.1 mL | |
9825 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
Z1P1F10*F12 | |
Western Blot, ELISA, Immunohistochemistry | |
KIAA0757FLJ13167, PD1spermatogenesis associated PD1, spermatogenesis associated 2, spermatogenesis-associated protein 2, Spermatogenesis-associated protein PD1, tamo | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title