Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570422
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
May have a role in the regulation of spermatogenesis.
Specifications
SPATA2 | |
Polyclonal | |
Unconjugated | |
SPATA2 | |
AI504642; KIAA0757; mKIAA0757; PD1; PPP1R145; protein phosphatase 1, regulatory subunit 145; SPATA2; spermatogenesis associated 2; spermatogenesis associated PD1; spermatogenesis-associated protein 2; spermatogenesis-associated protein PD1; tamo | |
Rabbit | |
Affinity chromatography | |
RUO | |
9825 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UM82 | |
SPATA2 | |
synthetic peptide directed towards the N-terminal region of human SPATA2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction