Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA25 Mouse anti-Human, Janelia Fluor 646, Clone: Z37P3H11*B2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP250305JF646
Description
SPATA25 Monoclonal antibody specifically detects SPATA25 in Human samples. It is validated for Immunohistochemistry.Specifications
SPATA25 | |
Z37P3H11&asterisk;B2 | |
Immunohistochemistry | |
Purified | |
SPATA25 | |
Peptide Sequence - LVRSKRGQP | |
Protein G purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunohistochemistry | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C20orf165, Chromosome 20 Open Reading Frame 165, DJ337O18.8, Spermatogenesis Associated 25, Testis-Specific Gene 23 Protein, TSG23 | |
Mouse | |
IgG | |
0.1mL | |
Primary | |
128497 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title