Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA5L1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550831

Catalog No. PA550831


Add to cart

Description

Description

The antibody detects endogenous levels of total SPATA5L1 protein.

The function of this protein remains unknown.
Specifications

Specifications

SPATA5L1
Polyclonal
Unconjugated
SPATA5L1
Liquid
SPATA5L1
Synthetic peptide corresponding to residues near the C terminal of human spermatogenesis associated 5-like
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9BVQ7
Spermatogenesis-associated protein 5-like protein 1
Rabbit
IgG
100 μL
-20°C
Primary
79029
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit