Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558241
Description
Immunogen sequence: RHLLHVLKVD LGCTSEENSV KQNDVDMLNV FDFEKAGNSE PNELKNESEV TIQQERQQYQ KALDMLLSAP KDENEIFPSP TEFFMPIYKS KHSEGV Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 55%.
The function of the SPATA7 protein remains unknown.
Specifications
SPATA7 | |
Polyclonal | |
Unconjugated | |
SPATA7 | |
AI661438; B230306G18Rik; epididymis secretory protein Li 296; fertility related protein WMP1; fertility-related protein WMP1; HEL-S-296; HSD3; HSD-3.1; LCA3; RSD-3; si:ch211-51o12.1; SPATA7; sperm DNA no.3; spermatogenesis associated 7; spermatogenesis-associated protein 7; Spermatogenesis-associated protein 7 homolog; spermatogenesis-associated protein 7 homolog; spermatogenesis-associated protein 7; spermatogenesis-associated protein HSD3; Wmp1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55812 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P0W8 | |
SPATA7 | |
Recombinant protein corresponding to Human SPATA7. Recombinant protein control fragment (Product #RP-95556). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction