Learn More
Aviva Systems Biology Corp SPATA9 Antibody - N-terminal region (ARP50025_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50025_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPATA9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPATA9. Alternate names: FLJ35906, NYD-SP16ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-62842