Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | SPATA9 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159840
|
Novus Biologicals
NBP159840 |
100 μL |
Each of 1 for $436.00
|
|
Description
SPATA9 Polyclonal specifically detects SPATA9 in Human samples. It is validated for Western Blot.Specifications
SPATA9 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ35906, NYD-SP16, spermatogenesis associated 9, spermatogenesis-associated protein 9, Testis development protein NYD-SP16 | |
SPATA9 | |
IgG | |
Affinity Purified | |
29 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9BWV2 | |
83890 | |
Synthetic peptides corresponding to SPATA9 (spermatogenesis associated 9) The peptide sequence was selected from the N terminal of SPATA9)(50ug). Peptide sequence FKDEFPTILRLSQSNQKREPAQKTSKIRMAIALAKINRATLIRGLNSISR. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title