Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATS2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549357

Catalog No. PA549357

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SPATS2
Polyclonal
PBS with 0.09% sodium azide
Q86XZ4
Spermatogenesis-associated serine-rich protein 2, Serine-rich spermatocytes and round spermatid 59 kDa protein, p59scr, SPATS2, SCR59, SPATA10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65244
Western Blot
Unconjugated
SPATS2
Liquid
SPATS2
KLH conjugated synthetic peptide between 448-482 amino acids from the C-terminal region of human SPATS2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit