Learn More
SPC Allwik™ SOCs Pillows
For use with all oil and water-based fluids
Manufacturer:  Sorbent Products Co Inc AW430
 View more versions of this product
Catalog No. 19-819-434
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
30 gal./case
3 in.
30 gal.
1.21m
Universal Sorbent Sock
Polypropylene
Gray
7.62cm
113.5L
4 ft.
Suitable for most industrial applications
30 gal., 113.5L