SPC Drum Top Covers

Manufacturer: Sorbent Products Co Inc DTA25

 View more versions of this product

Catalog No. 19-819-440

  Check Availability
Add to Cart

    Specifications

    Color Gray
    Diameter (English) Inner 22 in.
    Diameter (Metric) Inner 55.8cm
    For Use With (Application) Absorbs all liquids, prevent pump drips from reaching the floor and keep drum tops and drum storage areas clean
    For Use With (Equipment) 55 gal. Dimpled drums
    Includes 1 or 2 Bung holes
    Product Type Drum Pre-cut Top Cover

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit