Learn More
SPC™ High Traffic Series Sorbents
Made to withstand heavy foot and forklift traffic along with problem spill areas
Manufacturer:  Sorbent Products Co Inc HT303
 View more versions of this product
Catalog No. 19-819-45
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Gray
238.48L
300 ft.
Walkways, overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
30 in.
63 gal., 238.48L
63 gal.
91.4m
High Traffic Series Universal Sorbent Roll
2 ply
76.2cm
300 ft. x 30 in. (91.4m x 76.2cm)