SPC™ Universal Plus™ Sorbent Pad

Effective with hazardous and aggressive fluids

Manufacturer: Sorbent Products Co Inc SPCUN1113

 View more versions of this product

Catalog No. 19-003-205

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) Overspray leaks and drips, workstations, heavy clean up and wipe up , drums - liquid storage, outfalls and discharge ponds
    Width (English) 12 in.
    Capacity (English) 20 gal.
    Capacity (Metric) 75.7L
    Length (Metric) 30.48cm
    Length (English) 12 in.
    Product Type Universal Plus Sorbent Pad
    Thickness 3 ply
    Width (Metric) 30.48cm
    Capacity 20 gal., 75.7L
    Dimensions (L x W) 12 x 12 in. (30.48 x 30.48cm)
    Color Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit