Learn More
SpCas9 Mouse anti-Bacteria, Clone: 14B6, GenScript™
Mouse Monoclonal Antibody
Supplier: Genscript Corporation A0193640

Description
Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. Clustered regularly interspaced short palindromic repeats (CRISPR)/CRISPR-associated (Cas) protein 9 system provides a robust and multiplexable genome editing tool, enabling researchers to precisely manipulate specific genomic elements, and facilitating the elucidation of target gene function in biology and diseases. CRISPR/Cas9 genome editing allows for double-stranded DNA breaks at specific sequences to efficiently disrupt, excise, mutate, insert, or replace genes. To date, the Streptococcus pyogenes Cas9 (SpCas9) has been used broadly to achieve efficient genome editing in a variety of species and cell types. It is important that the precision of transfection and the level of Cas9 expression should be controlled strictly during the editing processes by using specific anti-CRISPR/Cas9 antibodies.
SpCas9 Monoclonal antibody specifically detects SpCas9 in Bacteria samples. It is validated for ELISA, Immunocytochemistry, Western BlotSpecifications
SpCas9 | |
Monoclonal | |
Unconjugated | |
cas9; CASP 9; Caspase9; Caspase-9 precursor; CASPASE-9c; CRISPR9; CRISPR-Cas9; CTD; OTTHUMP00000002324; RNCASP9; RP11-265F14.3; s aureus | |
Recombinant Streptococcus pyogenes CRISPR/Cas9 | |
40 μg | |
Primary | |
-20°C or -80°C if preferred | |
Lyophilized |
ELISA, Immunocytochemistry, Western Blot | |
14B6 | |
PBS with 0.02% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.