Learn More
Starna Cells Inc SPCTROSL FAR UV FLW CELL STND
SDP
Supplier:  Starna Cells Inc 71Q10
Encompass_Preferred
Spectrosil FAR UV Quartz Flow Cell standard 10mm Useable range: 170 to 2700 nm
Catalog No. NC1312787