Learn More
Aviva Systems Biology Corp SPDYA Antibody - middle region (ARP55846_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55846_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPDYA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPDYA. Alternate names: MGC110856, MGC57218, SPDY1, SPY1, RINGO3, RINGOAApplicationWB. Species reactivity: Human
Catalog No. 50-014-61633