Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPDYE1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548165

Catalog No. PA548165

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is located at chromosome 7p13 which is close to the Williams Beuren syndrome chromosome region 7q11.23. [provided by RefSeq, Jul 2008]
Specifications

Specifications

SPDYE1
Polyclonal
PBS with 0.09% sodium azide
Q8NFV5
Speedy protein E1, Williams-Beuren syndrome chromosomal region 19 protein, SPDYE1, WBSCR19
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
285955
Flow Cytometry, Western Blot
Unconjugated
SPDYE1
Liquid
SPDYE1
KLH conjugated synthetic peptide between 242-271 amino acids from the C-terminal region of human SPDYE1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit