Learn More
Speakman™ Eyesaver Emergency Eyewash
Manufacturer:  Speakman™ SE580
Catalog No. 19-069132
Add to cart
Specifications
Specifications
Eyewash
Triangular pull handle, Stainless Steel push handle
3.6 gpm at 30psi
Floor
Safety
ANSI Z358.1
Ball valves, aerated spray outlets, flip top dust covers, deluge shower head, yellow ABS Plastic bowl, Chrome Plated valve