Learn More
Speakman™ Eyesaver Free-Standing Eye/Face Washes
Manufacturer:  Speakman™ SE420
Catalog No. 19-058-402
Add to cart
Specifications
Specifications
Eye/Facewash
Stainless Steel push handle
Stainless Steel rectangular bowl, six aerated sprays, stay open ball valve, in-line strainer, yellow Polypropylene shroud, IPS Galvanized Steel pipe
1.25 in. NPT female
Safety
ANSI Z358.1
Pedestal