Speakman™ Eyesaver™ Wall-Mounted Eye/Face Washes
Wall-mounted emergency eyewash
Manufacturer: Speakman CompanySupplier Diversity Partner SE505
 View more versions of this product
Catalog No. 19-039-602
  Check Availability
Add to Cart
    Specifications
    Product Type Eyewash
    Flow Rate 3.6 gpm at 30psi
    For Use With (Application) Rinsing of the eyes and face
    Includes Twin aerated yellow Plastic spray head, rectangular Stainless Steel bowl, Chrome Plated Brass NTP, universal emergency sign
    Mounting Wall
    Activation Stainless Steel trigger bar
    Certifications/Compliance ANSI Z358.1
  • Description & Specifications
Related Products
Product Content Feedback