Learn More
Speakman™ Vertical Deluge Shower
Manufacturer:  Speakman™ SE220
Catalog No. 19-819-511
Add to cart
Specifications
Specifications
Emergency Shower
ANSI Z358.1
Yellow Plastic shower head, NPT female Brass, Chrome Plated stay open valve, universal emergency sign
Powder coated yellow Aluminum triangular pull rod
20 gpm
30psi