Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spearmint Oil, Natural, FCC, 55%, Spectrum™

Click to view available options
Quantity:
100 mL
2.5 L
200 L
25 lbs.
50 kg
500 mL
Packaging:
Amber Glass Bottle
Drum
Steel Drum
Steel Pail
8008-79-5
8008-79-5

SP306, 8008-79-5

Spectrum™ Spearmint Oil, Natural, FCC is used for cooking, flavoring, and as a digestive support. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Steel Drum
Quantity 50 kg
Refractive Index 1.484-1.491
CAS Max % 1
Percent Purity 55%
Grade FCC