Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPECC1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564448
Description
Immunogen sequence: QRHSISGPIS TSKPLTALSD KRPNYGEIPV QEHLLRTSSA SRPASLPRVP AMESAKT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
This gene encodes a protein that is leucine-rich and is thought to play a role in regulating the interaction of the cytoskeleton with a variety of cellular processes. Mutations in this gene are associated with the French-Canadian type of Leigh syndrome. Transcripts ranging in size from 4. 8 to 7. 0 kb which result from alternative polyadenylation have been reported for this gene.
Specifications
SPECC1L | |
Polyclonal | |
Unconjugated | |
SPECC1L | |
4930470P14Rik; 4932439K10Rik; 9530057A13Rik; AI317249; cytokinesis and spindle organization A; cytospin A; cytospin-A; CYTSA; GBBB2; Kiaa0376; mKIAA0376; OBLFC1; Renal carcinoma antigen NY-REN-22; RGD1309570; Specc1l; SPECC1-like; SPECC1-like protein; sperm antigen with calponin homology and coiled-coil domains 1 like; sperm antigen with calponin homology and coiled-coil domains 1-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23384 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q69YQ0 | |
SPECC1L | |
Recombinant protein corresponding to Human SPECC1L. Recombinant protein control fragment (Product #RP-104118). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction