Learn More
Tocris Bioscience™ U 104 Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Tocris Bioscience™ 4540/10
 View more versions of this product
Catalog No. 45-401-0
Add to cart
This item is not returnable. View return policy
Description
Description
Potent carbonic anhydrase (CA) inhibitor (Ki values are 4.5, 45.1, 5080 and 9640 nM for CA XII, CA IX, CA I and CA II respectively in MDA-MB-231 cells). Inhibits tumor metastasis in the rat 4T1 metastasis model.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
4-[[[(4-Fluorophenyl)amino]carbonyl]amino]-benzesulfonamide
>99%
Observed MW: 309.32
Soluble to 100mM in DMSO
178606-66-1
C13H12FN3O3S
10mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT