Learn More
Tocris Bioscience™ U 104
Manufacturer:  Tocris Bioscience™ 4540/50
 View more versions of this product
Catalog No. 45-405-0
Add to cart
This item is not returnable. View return policy
Description
Description
Potent carbonic anhydrase (CA) inhibitor (Ki values are 4.5, 45.1, 5080 and 9640 nM for CA XII, CA IX, CA I and CA II respectively in MDA-MB-231 cells). Inhibits tumor metastasis in the rat 4T1 metastasis model.
Specifications
Specifications
4-[[[(4-Fluorophenyl)amino]carbonyl]amino]-benzesulfonamide
>99%
Observed MW: 309.32
Soluble to 100mM in DMSO
178606-66-1
C13H12FN3O3S
50mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT