Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Specific Gravity Standard, 1.17 at 25°C/25°C, Reagents

Catalog No. 01333484
Click to view available options
Quantity:
500 mL
1 L

Specific Gravity Standard, 1.17 at 25°C/25°C, Reagent, use for Specific gravity calibration. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Specific Gravity Standard, 1.17 at 25°C/25°C
Packaging Poly Bottle
CAS 7732-18-5, 7647-15-6
Quantity 500 mL
Specific Gravity 1.17
Physical Form Liquid
Color Colorless
pH 6.5 to 8
Boiling Point 102°C
Solubility Completely miscible
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.