Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Specimen Pails with Cover
SDPSCI_ED

Heavy-duty HDPE pails.

Supplier:  Dynalon™ 413045

Catalog No. S30518A

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.

This item is not returnable. View return policy

Description

Description

  • Molded, high-density polyethylene
  • Rigid construction
  • Steel bail handle with plastic hand grip
  • Snap-on cover
  • Capacity: 12 qt. (11L)

Specifications

Specifications

Specimen Pail
HDPE
White
9.75 in.
steel bail carrying handle with plastic hand grip and snap-on cover
29.21 cm
11 L
2.4 gal.
Steel
24.76 cm
11.5 in.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit