Learn More
Epredia™ Specimen Stages for HM 520, 525, 550 and 560
Use Epredia™ Specimen Stages for Epredia™ HM 520, 525, 550 and 560 Series with HM 520, 525, 550 and 560 Cryostats..
Manufacturer:  Epredia™ 715730
 View more versions of this product
Catalog No. 71-573-0
Add to cart
Description
Description
Specifications
Specifications
50 x 50mm
For HM 550 Series
Specimen Stages
For HM 550 Series