Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectinomycin dihydrochloride pentahydrate, 50mg/mL in water, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
22189-32-8
C14H36Cl2N2O12
495.344
MFCD00150886
DCHJOVNPPSBWHK-UXXUFHFZSA-N
67108398
CHEBI:9217
[H+].[H+].CC1CC(=O)C2(C(O1)OC3C(C(C(C(C3O2)NC)O)NC)O)O.O.O.O.O.O.[Cl-].[Cl-]
22189-32-8
C14H36Cl2N2O12
495.344
MFCD00150886
DCHJOVNPPSBWHK-UXXUFHFZSA-N
67108398
CHEBI:9217
[H+].[H+].CC1CC(=O)C2(C(O1)OC3C(C(C(C(C3O2)NC)O)NC)O)O.O.O.O.O.O.[Cl-].[Cl-]
Applications
It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Spectinomycin dihydrochloride pentahydrate, 50mg/mL in water

RTECSNumber : WG7400000

storageNote1 : -30° to -10°C