Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Spectra™ Multicolor Broad Range Protein Ladder

A 4-color protein standard containing 10 prestained proteins (10 to 260kDa) for use as size standards in gel electrophoresis and Western blotting.

Supplier:  Thermo Scientific™ 26634

 View more versions of this product

Catalog No. PI26634


Add to Cart

Description

Description

Thermo Scientific™ Spectra Multicolor Broad Range Protein Ladder is a 4-color protein standard containing 10 prestained proteins (10 to 260kDa) for use as size standards in gel electrophoresis and Western blotting.

This prestained protein MW marker is designed for monitoring the progress of SDS-polyacrylamide gel electrophoresis, for assessing transfer efficiency onto PVDF, nylon and nitrocellulose membranes, and for estimating the approximate size of separated proteins that have been made visible with gel stains or Western blot detection reagents. Four different chromophores (blue, orange, green, pink) are bound to the different component proteins, producing a brightly colored ladder with an easy-to-remember pattern.

Highlights:

  • Size range – 10 proteins spanning 10 to 260kDa

  • Multicolor – four different colors for unambiguous band-size assignment
  • Ready-to-use – supplied in a loading buffer for direct loading on gels; no need to boil
  • Sharp bands – color-coded bands of similar intensity for easy visualization
  • Quality tested – each lot evaluated by SDS-PAGE and Western blotting
  • Membrane-compatible – colored bands transfer to membranes for Western blotting

Includes:

Dye-stained proteins in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 10mM DTT, 1mM NaN3 and 33% glycerol.

Recommended for:

Monitoring protein migration during SDS-polyacrylamide gel electrophoresis; Monitoring protein transfer onto membranes after Western blotting; Sizing of proteins on SDS-polyacrylamide gels and Western blots

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contents: two vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

10
Yes
4 colors: Blue, Orange, Green, Pink
260, 140, 100, 70, 50, 40, 35, 25, 15, 10 kDa
2 x 250 μL
Western Blotting, SDS-PAGE
Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Protein Ladder
10 to 260 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Spectra™
Approved for shipment on Wet or Dry Ice
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.