Learn More
Thermo Scientific™ Spectra™ Multicolor Low Range Protein Ladder
A mixture of six proteins and synthetic peptides (1.7 to 40kDa) for use as size standards for peptides and small proteins in gel electrophoresis and Western blotting.
Specifications
Description Spectra Multicolor Low Range Protein Ladder
Detection Visible
Formulation 6 peptides and proteins from 1.7 to 40kDa in reducing sample buffer
Stained Prestained
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI26629
View Documents
Thermo Scientific™
26629
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description
Description
Thermo Scientific™ Spectra Multicolor Low Range Protein Ladder is a mixture of six (6) proteins and synthetic peptides (1.7 to 40kDa) for use as size standards for peptides and small proteins in gel electrophoresis and Western blotting.
This prestained protein MW marker is designed for monitoring the progress of SDS-polyacrylamide gel electrophoresis, for assessing transfer efficiency onto PVDF, nylon and nitrocellulose membranes, and for estimating the approximate size of separated proteins that have been made visible with gel stains or Western blot detection reagents. Three different chromophores (blue, orange, green) are bound to the different component proteins, producing a brightly colored ladder with an easy-to-remember pattern. The Spectra Multicolor High Range Protein Ladder is ready to use: no heating, further dilution or addition of a reducing agent is required.
Highlights:
  • Size range – six proteins and peptides spanning 1.7 to 40kDa
  • Multicolor – three different colors for unambiguous band-size assignment
  • Ready-to-use – supplied in a loading buffer for direct loading on gels; no need to boil
  • Sharp bands – color-coded bands of similar intensity for easy visualization
  • Quality tested – each lot evaluated by SDS-PAGE and Western blotting
  • Membrane-compatible – colored bands transfer to membranes for Western blotting
Includes:
Dye-stained proteins in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 10mM DTT, 1mM NaN3 and 33% glycerol.
Recommended for:
Monitoring of protein migration during SDS-PAGE; Verifying Western transfer efficiency; Approximate sizing of proteins on SDS-polyacrylamide gels and Western blots; Locating a protein of interest for excision from an unstained preparative gel
Specifications
Specifications
Spectra Multicolor Low Range Protein Ladder
Visible
6 peptides and proteins from 1.7 to 40kDa in reducing sample buffer
Prestained