Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin beta 3 Mouse anti-Human, PE/Atto594, Clone: SPTBN2/1583, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254440PEATT594

Catalog No. N254440P594


Add to cart

Description

Description

Spectrin beta 3 Monoclonal antibody specifically detects Spectrin beta 3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Spectrin beta 3
SPTBN2/1583
Flow Cytometry
Conjugated, Purified
SPTBN2
Recombinant human SPTBN2 fragment (aa356-475) (exact sequence is proprietary)
Protein A or G purified
Neuroscience
Primary
6712.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Beta-III spectrin, glutamate transporter EAAT4-associated protein 41, GTRAP41, KIAA0302, SCA5, spectrin beta chain, brain 2, spectrin, beta, non-erythrocytic 2, Spectrin, non-erythroid beta chain 2, spinocerebellar ataxia 5
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit