Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin beta 3 Rabbit anti-Human, Mouse, Clone: 1287A, Novus Biologicals™

Rabbit MonoclonalAntibody

$99.70 - $300.50

Specifications

Antigen Spectrin beta 3
Clone 1287A
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 5 - 20 ug/ml, Immunohistochemistry-Paraffin 1:1000, Flow (Intracellular) 1 ug/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
NBP259668 View Documents Novus BiologicalsSupplier Diversity Partner
NBP259668
0.1 mg Unconjugated Each for $300.50
Add to cart
 
NBP259668SS View Documents Novus BiologicalsSupplier Diversity Partner
NBP259668SS
0.025 mg Unconjugated Each for $99.70
Add to cart
 
Description

Description

Spectrin beta 3 Monoclonal antibody specifically detects Spectrin beta 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications

Specifications

Spectrin beta 3
Western Blot 2 ug/ml, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 5 - 20 ug/ml, Immunohistochemistry-Paraffin 1:1000, Flow (Intracellular) 1 ug/mL
Purified
Human, Mouse
PBS with No Preservative
Synthetic peptide containing the C-terminal region of human Spectrin beta 3 (UniProt O15020)
Monoclonal
Protein A or G purified
1287A
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Rabbit
RUO
6712
IgG
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit