Specifications
Shorwave UV tube w/ Longlife glass filter | |
Mineralogy, stamp collecting, microbiology, mutation studies, laboratory UV dosing and illumination | |
1.25 in. | |
5 W |
254 nm | |
9 in. | |
3.17 cm |