Learn More
Spectroline™ Pencil Shortwave/Longwave UV Lamps
Manufacturer:  Spectroline™ 11SC1
 View more versions of this product
Catalog No. 11-992-30
Add to cart
Requires : Power supply for lamp sold separately
Description
Description
Lightweight and miniature in size. Ideal for insertion into small apertures requiring spectroscopic wavelength calibration, sterilization, UV curing, or fluorescence detection applications. These low-pressure, mercury-vapor discharge lamps maintain a high output up to 5000 hours. Double-bore quartz construction with electrodes located at one end for easy installation.
†Relative intensity at 1 in. (2.54cm).
Specifications
Specifications
0.25 in.
2.12 in.
UV pencil lamp
254nm
6.35mm
53.9mm
Each