Learn More
Spectroline™ Pencil Shortwave/Longwave UV Lamps
Manufacturer:  Spectroline™ 36380
 View more versions of this product
Catalog No. 11-992-55
Add to cart
Requires : Power supply for lamp sold separately
Description
Description
Lightweight and miniature in size. Ideal for insertion into small apertures requiring spectroscopic wavelength calibration, sterilization, UV curing, or fluorescence detection applications. These low-pressure, mercury-vapor discharge lamps maintain a high output up to 5000 hours. Double-bore quartz construction with electrodes located at one end for easy installation.
†Relative intensity at 1 in. (2.54cm).
Specifications
Specifications
Each
0.375 in.
2.12 in.
UV pencil lamp
365nm
9.52mm
53.9mm