Learn More
Spectroline Replacement Tubes for UV Cabinets FB-PDS-150, -151
Manufacturer: Spectroline BLE1T151
Specifications
15w | |
Replacement Tube | |
600μw/cm2 |
365nm | |
UV cabinet FB-PDS-150, -151 |