Spectroline Replacement Tubes
Manufacturer: Spectronics CorporationSupplier Diversity Partner BLE6T312
 View more versions of this product
Catalog No. 11-992-105
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) B series battery operated UV hand lamp
    Intensity 620μw/cm2
    Wattage 6w
    Product Type Replacement Tube
    Wavelength 312nm
  • Description & Specifications
Related Products
Product Content Feedback