Learn More
Spectronics Corporation Hinged UV-Blocking Cover for Spectroline Slimline UV Transilluminators
Manufacturer:  Spectronics CorporationSupplier Diversity Partner UVC250A
 View more versions of this product
Catalog No. 11-993-08
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1 in.
18 in.
TC, TD, TL, TR, TS and TV series transilluminator
13 in.
45.7cm
UV Transmitting Filter Protector
Allows manipulation of gels, prevents damage to the filter, resistant to solarization
2.5cm
33cm
13 x 18 x 1 in. (33 x 45.7 x 2.5cm)