Spectronics Corporation Ultraviolet Viewing Cabinets: Darkroom

Designed for viewing electrophoresis gels and TLC plates

Manufacturer: Spectronics CorporationSupplier Diversity Partner CX20

 View more versions of this product

Catalog No. 05-720-30

  Check Availability
Add to Cart

    Specifications

    Product Type Darkroom cabinet with soft rubber curtains
    Width (English) 8.7 in.
    For Use With (Application) UV viewing
    Includes Contoured eyepiece; Soft rubber curtain; Removable bottom panel; 8w UV-A, UV-C tubes
    Height (Metric) 22.9cm
    Wavelength 365/254nm
    Width (Metric) 22.9cm
    Height (English) 9 in.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit