Learn More
Spectroline™ Transmitting Filter Protector for UV Transilluminators
Manufacturer:  Spectroline™ UVT150A
Catalog No. 11-993-3
Add to cart
Description
Description
  • For use with variable-intensity transilluminator 11-992-83 or dual UV/white-light transilluminator 11-992-81
Specifications
Specifications
0.4 in.
19.25 in.
TC, TD, TL, TR, TS and TV series transilluminator
12.2 in.
48.89cm
UV Transmitting Filter Protector
Allows manipulation of gels, prevents damage to the filter, resistant to solarization
1cm
31cm
12.2 x 19.25 x 0.4 in. (31 x 48.89 x 1cm)