Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum Brands Rayovac Electronic Batteries

Manufacturer:  Spectrum Brands KECR20251

Catalog No. 19-120-2753

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

2025
No
Pressure contact
0.002 lb.
Coin cell
0.78 in.
Lithium Manganese Dioxide
3V
Lithium Battery
1.2g
20mm
For watches, calculators, remote controls and medical devices
165mAH
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit