Learn More
Spectrum™ Chromatography Aqueous Chromatography Columns
Manufacturer:  Spectrum ChromatographySupplier Diversity Partner 124021
 View more versions of this product
Catalog No. 11-190-207
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Standard Buffers for high quality, classic chromatographic separation
40psi
200mm
25mm
Aqueous Chromatography Column
98mL
Borosilicate Glass
0.18 in. I.D. Tubing connectors