Learn More
Spectrum™ Chromatography Aqueous Chromatography Columns
Manufacturer:  Spectrum ChromatographySupplier Diversity Partner 124014
 View more versions of this product
Catalog No. 22-265216
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Standard Buffers for high quality, classic chromatographic separation
45psi
600mm
15mm
Aqueous Chromatography Column
106mL
Borosilicate Glass
0.18 in. I.D. Tubing connectors