Spectrum™ Chromatography Spectra/Chrom™ Aqueous Chromatography Columns
Manufacturer: Spectrum ChromatographySupplier Diversity Partner 123945
 View more versions of this product
Catalog No. 11-190-189
  Check Availability
Add to Cart
    Specifications
    Type Spectra/Chrom™ Aqueous chromatography column
    Length (Metric) 100cm
    Diameter (Metric) Inner 6mm
    Includes 10μm bed support for use with most low-pressure media 0.1875 in. O.D. tube fittings, Polypropylene endplates
    For Use With Aqueous solutions including most Acids, bases, and Salts
    Max. Pressure 50psi
    Material Fluoropolymer Resin, Polypropylene, Borosilicate Glass, and Nitrile Rubber
  • Description & Specifications
Related Products
Product Content Feedback