Learn More
Spectrum™ Chromatography Spectra/Chrom™ Aqueous Chromatography Columns
Manufacturer:  Spectrum ChromatographySupplier Diversity Partner 124017
 View more versions of this product
Catalog No. 11-190-206
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Aqueous solutions including most Acids, bases, and Salts
Fluoropolymer Resin, Polypropylene, Borosilicate Glass, and Nitrile Rubber
80cm
10μm bed support for use with most low-pressure media 0.1875 in. O.D. tube fittings, Polypropylene endplates
45psi
Spectra/Chrom™ Aqueous chromatography column
15mm