Spectrum™ Chromatography Spectra/Chrom™ Aqueous Chromatography Columns
Manufacturer: Spectrum ChromatographySupplier Diversity Partner 124044
 View more versions of this product
Catalog No. 06-651-5E
  Check Availability
Add to Cart
    Specifications
    For Use With Aqueous solutions including most Acids, bases, and Salts
    Type Spectra/Chrom™ Aqueous chromatography column
    Length (Metric) 150cm
    Diameter (Metric) Inner 50mm
    Includes 10μm bed support for use with most low-pressure media 0.25 in. O.D. tube fittings, Polypropylene endplates
    Max. Pressure 25psi
    Material Fluoropolymer Resin, Polypropylene, Borosilicate Glass, and Nitrile Rubber
  • Description & Specifications
Related Products
Product Content Feedback